Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOOK1
Entrez Gene ID
51361
Gene Name
hook microtubule-tethering protein 1
Gene Aliases
HK1
Chromosome Location
Chr.1: 59814861 - 59876378 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015888.4 NP_056972.1 20 - 21 2200 145
XM_011541562.2 XP_011539864.1 20 - 21 2170 145
GenBank mRNA AF044923.1 - 20 - 21 2142 145
AK097735.1 - 18 - 19 1943 145
AK098561.1 - 14 - 15 1481 145
AK292314.1 - 21 - 22 2045 145
AK301768.1 - 20 - 21 1968 145
BC011621.2 - 20 - 21 2200 145

View Full Product Details