The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015888.4 | NP_056972.1 | 7 - 8 | 786 | 68 |
XM_006710676.1 | XP_006710739.1 | 7 - 8 | 726 | 68 | |
XM_011541562.2 | XP_011539864.1 | 7 - 8 | 756 | 68 | |
XM_011541563.1 | XP_011539865.1 | 7 - 8 | 726 | 68 | |
XM_017001424.1 | XP_016856913.1 | 7 - 8 | 726 | 68 | |
GenBank mRNA | AF044923.1 | - | 7 - 8 | 728 | 68 |
AK097735.1 | - | 7 - 8 | 647 | 68 | |
AK098561.1 | - | 3 - 4 | 184 | 68 | |
AK292314.1 | - | 8 - 9 | 631 | 68 | |
AK301768.1 | - | 7 - 8 | 554 | 68 | |
BC011621.2 | - | 7 - 8 | 786 | 68 | |
BC107857.1 | - | 7 - 8 | 700 | 68 |