The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006842.2 | NP_006833.2 | 16 - 17 | 2013 | 61 |
XM_005273726.3 | XP_005273783.1 | 16 - 17 | 2583 | 61 | |
XM_011544740.2 | XP_011543042.1 | 16 - 17 | 2583 | 61 | |
XM_017017144.1 | XP_016872633.1 | 16 - 17 | 2580 | 61 | |
GenBank mRNA | AK290850.1 | - | 16 - 17 | 1997 | 61 |
AK300016.1 | - | 10 - 11 | 1287 | 61 | |
BC000401.2 | - | 16 - 17 | 1972 | 61 | |
BC007610.1 | - | 9 - 10 | 1196 | 61 | |
BC014125.2 | - | 4 - 5 | 401 | 61 | |
BC053577.1 | - | 16 - 17 | 1980 | 61 | |
BX537771.1 | - | 14 - 15 | 1686 | 61 | |
U41371.1 | - | 16 - 17 | 1952 | 61 |