The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006842.2 | NP_006833.2 | 20 - 21 | 2467 | 135 |
XM_005273726.3 | XP_005273783.1 | 20 - 21 | 3037 | 135 | |
XM_011544740.2 | XP_011543042.1 | 20 - 21 | 3037 | 135 | |
XM_017017144.1 | XP_016872633.1 | 20 - 21 | 3034 | 135 | |
GenBank mRNA | AK126488.1 | - | 1 - 2 | 3402 | 135 |
AK290850.1 | - | 20 - 21 | 2451 | 135 | |
AK300016.1 | - | 14 - 15 | 1741 | 135 | |
BC000401.2 | - | 20 - 21 | 2426 | 135 | |
BC007610.1 | - | 13 - 14 | 1650 | 135 | |
BC014125.2 | - | 8 - 9 | 855 | 135 | |
BC053577.1 | - | 20 - 21 | 2434 | 135 | |
BE677116.1 | - | 2 - 3 | 135 | ||
BX537771.1 | - | 18 - 19 | 2140 | 135 | |
U41371.1 | - | 20 - 21 | 2406 | 135 |