Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EHBP1L1
Entrez Gene ID
254102
Gene Name
EH domain binding protein 1 like 1
Gene Aliases
-
Chromosome Location
Chr.11: 65576034 - 65592650 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099409.1 NP_001092879.1 2 - 3 430 67
XM_005273867.3 XP_005273924.1 2 - 3 433 67
XM_005273868.3 XP_005273925.1 2 - 3 432 67
XM_005273869.3 XP_005273926.1 2 - 3 434 67
XM_005273870.3 XP_005273927.1 2 - 3 433 67
XM_006718486.3 XP_006718549.1 2 - 3 431 67
XM_006718487.3 XP_006718550.1 2 - 3 431 67
XM_017017469.1 XP_016872958.1 2 - 3 432 67
GenBank mRNA AL834433.1 - 2 - 3 127 67
BX443367.2 - 2 - 3 430 67

View Full Product Details