The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139159.4 | NP_631898.3 | 16 - 17 | 2149 | 70 |
XM_005259673.2 | XP_005259730.1 | 17 - 18 | 2258 | 70 | |
XM_011528405.1 | XP_011526707.1 | 18 - 19 | 2310 | 70 | |
XM_011528406.1 | XP_011526708.1 | 19 - 20 | 2420 | 70 | |
XM_011528408.1 | XP_011526710.1 | 16 - 17 | 2095 | 70 | |
XM_011528410.1 | XP_011526712.1 | 17 - 18 | 2310 | 70 | |
XM_017027453.1 | XP_016882942.1 | 18 - 19 | 2900 | 70 | |
XM_017027454.1 | XP_016882943.1 | 19 - 20 | 3010 | 70 | |
GenBank mRNA | AF452102.2 | - | 16 - 17 | 2169 | 70 |
AF542510.2 | - | 16 - 17 | 2128 | 70 | |
AK054656.1 | - | 5 - 6 | 689 | 70 | |
AK122645.1 | - | 10 - 11 | 1199 | 70 | |
AK122654.1 | - | 16 - 17 | 2152 | 70 | |
AK131100.1 | - | 13 - 14 | 1718 | 70 | |
AK131499.1 | - | 15 - 16 | 1662 | 70 | |
AL834376.1 | - | 10 - 11 | 1180 | 70 | |
AY172660.1 | - | 13 - 14 | 1814 | 70 | |
AY374518.1 | - | 13 - 14 | 1804 | 70 | |
BC000970.1 | - | 4 - 5 | 532 | 70 | |
BC037948.2 | - | 16 - 17 | 2086 | 70 | |
BF203783.1 | - | - | 286 | 70 | |
CR627380.1 | - | 8 - 9 | 924 | 70 | |
DQ417928.1 | - | 14 - 15 | 1891 | 70 |