Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPP9
Entrez Gene ID
91039
Gene Name
dipeptidyl peptidase 9
Gene Aliases
DP9, DPLP9, DPRP-2, DPRP2
Chromosome Location
Chr.19: 4675226 - 4723843 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139159.4 NP_631898.3 3 - 4 316 70
XM_005259673.2 XP_005259730.1 4 - 5 425 70
XM_011528408.1 XP_011526710.1 3 - 4 262 70
GenBank mRNA AF452102.2 - 3 - 4 336 70
AF542510.2 - 3 - 4 295 70
AK122654.1 - 3 - 4 319 70
BC037948.2 - 3 - 4 253 70
DB105598.1 - 3 - 4 316 70
DQ417928.1 - 1 - 2 58 70

View Full Product Details