The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037175.2 | NP_001032252.1 | 1 - 2 | 148 | 56 |
NM_017982.3 | NP_060452.3 | 1 - 2 | 148 | 56 | |
XM_005273172.1 | XP_005273229.1 | 1 - 2 | 73 | 56 | |
XM_006711408.1 | XP_006711471.1 | 1 - 2 | 146 | 56 | |
XM_011509687.1 | XP_011507989.1 | 1 - 2 | 73 | 56 | |
XM_017001583.1 | XP_016857072.1 | 1 - 2 | 182 | 56 | |
XM_017001584.1 | XP_016857073.1 | 1 - 2 | 181 | 56 | |
XM_017001589.1 | XP_016857078.1 | 1 - 2 | 186 | 56 | |
GenBank mRNA | AK295553.1 | - | 1 - 2 | 111 | 56 |
AK296923.1 | - | 1 - 2 | 73 | 56 | |
AY358495.1 | - | 1 - 2 | 125 | 56 | |
DA125047.1 | - | 1 - 2 | 153 | 56 | |
EF560748.1 | - | 1 - 2 | 76 | 56 |