Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOL10
Entrez Gene ID
79954
Gene Name
nucleolar protein 10
Gene Aliases
PQBP5
Chromosome Location
Chr.2: 10570766 - 10689991 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261394.1 NP_001248323.1 10 - 11 874 66
GenBank mRNA AK024137.1 - 10 - 11 859 66

View Full Product Details