The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261392.1 | NP_001248321.1 | 16 - 17 | 1460 | 111 |
NM_001261394.1 | NP_001248323.1 | 16 - 17 | 1388 | 111 | |
NM_024894.3 | NP_079170.2 | 17 - 18 | 1538 | 111 | |
NR_048552.1 | 16 - 17 | 1492 | 111 | ||
XM_011510398.2 | XP_011508700.1 | 16 - 17 | 1405 | 111 | |
XM_011510399.2 | XP_011508701.1 | 15 - 16 | 1343 | 111 | |
XM_017004990.1 | XP_016860479.1 | 13 - 14 | 1063 | 111 | |
XM_017004991.1 | XP_016860480.1 | 6 - 7 | 515 | 111 | |
GenBank mRNA | AK290680.1 | - | 17 - 18 | 1502 | 111 |
AK297169.1 | - | 16 - 17 | 1448 | 111 | |
AK299617.1 | - | 16 - 17 | 1480 | 111 | |
BC005125.2 | - | 17 - 18 | 1518 | 111 | |
BM021448.1 | - | 4 - 5 | 302 | 111 | |
DN602355.1 | - | 2 - 3 | 166 | 111 |