The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261392.1 | NP_001248321.1 | 6 - 7 | 583 | 118 |
NM_001261394.1 | NP_001248323.1 | 7 - 8 | 661 | 118 | |
NM_024894.3 | NP_079170.2 | 7 - 8 | 661 | 118 | |
NR_048552.1 | 6 - 7 | 615 | 118 | ||
XM_011510400.2 | XP_011508702.1 | 7 - 8 | 649 | 118 | |
XM_011510402.2 | XP_011508704.1 | 7 - 8 | 649 | 118 | |
XM_017004990.1 | XP_016860479.1 | 5 - 6 | 403 | 118 | |
GenBank mRNA | AK024137.1 | - | 7 - 8 | 646 | 118 |
AK290680.1 | - | 7 - 8 | 625 | 118 | |
AK297169.1 | - | 6 - 7 | 571 | 118 | |
AK299617.1 | - | 6 - 7 | 603 | 118 | |
BC005125.2 | - | 7 - 8 | 641 | 118 |