Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STK33
Entrez Gene ID
65975
Gene Name
serine/threonine kinase 33
Gene Aliases
-
Chromosome Location
Chr.11: 8335029 - 8594289 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289058.1 NP_001275987.1 4 - 5 501 153
NM_001289061.1 NP_001275990.1 3 - 4 766 153
NM_030906.3 NP_112168.1 4 - 5 915 153
XM_011520288.2 XP_011518590.1 4 - 5 1993 153
XM_011520290.2 XP_011518592.1 6 - 7 912 153
XM_011520292.2 XP_011518594.1 4 - 5 714 153
XM_011520293.2 XP_011518595.1 4 - 5 743 153
XM_011520294.1 XP_011518596.1 3 - 4 708 153
XM_011520295.2 XP_011518597.1 3 - 4 2804 153
XM_011520297.2 XP_011518599.1 3 - 4 481 153
XM_017018143.1 XP_016873632.1 6 - 7 972 153
XM_017018144.1 XP_016873633.1 6 - 7 874 153
XM_017018145.1 XP_016873634.1 5 - 6 2906 153
XM_017018146.1 XP_016873635.1 6 - 7 1036 153
XM_017018147.1 XP_016873636.1 5 - 6 2833 153
XM_017018148.1 XP_016873637.1 4 - 5 2765 153
XM_017018149.1 XP_016873638.1 5 - 6 917 153
XM_017018150.1 XP_016873639.1 5 - 6 772 153
XM_017018151.1 XP_016873640.1 5 - 6 731 153
XM_017018152.1 XP_016873641.1 3 - 4 1062 153
XM_017018153.1 XP_016873642.1 3 - 4 660 153
XM_017018154.1 XP_016873643.1 3 - 4 519 153
XM_017018155.1 XP_016873644.1 3 - 4 1074 153
XM_017018156.1 XP_016873645.1 3 - 4 520 153
XM_017018157.1 XP_016873646.1 3 - 4 285 153
XM_017018158.1 XP_016873647.1 4 - 5 571 153
XM_017018159.1 XP_016873648.1 5 - 6 2909 153
XM_017018160.1 XP_016873649.1 3 - 4 708 153
XM_017018161.1 XP_016873650.1 5 - 6 2915 153
XM_017018162.1 XP_016873651.1 4 - 5 518 153
XM_017018163.1 XP_016873652.1 3 - 4 509 153
XM_017018164.1 XP_016873653.1 3 - 4 395 153
XM_017018165.1 XP_016873654.1 4 - 5 474 153
GenBank mRNA AJ303380.2 - 2 - 3 1259 153
AK093251.1 - 3 - 4 624 153
AK293193.1 - 4 - 5 367 153
BC031231.1 - 4 - 5 863 153

View Full Product Details