Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STK33
Entrez Gene ID
65975
Gene Name
serine/threonine kinase 33
Gene Aliases
-
Chromosome Location
Chr.11: 8335029 - 8594289 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289058.1 NP_001275987.1 6 - 7 719 105
NM_001289059.1 NP_001275988.1 3 - 4 426 105
NM_001289061.1 NP_001275990.1 5 - 6 984 105
NM_030906.3 NP_112168.1 6 - 7 1133 105
XM_011520288.2 XP_011518590.1 6 - 7 2211 105
XM_011520290.2 XP_011518592.1 8 - 9 1130 105
XM_011520292.2 XP_011518594.1 6 - 7 932 105
XM_011520293.2 XP_011518595.1 6 - 7 961 105
XM_011520294.1 XP_011518596.1 5 - 6 926 105
XM_011520295.2 XP_011518597.1 5 - 6 3022 105
XM_011520297.2 XP_011518599.1 5 - 6 699 105
XM_017018143.1 XP_016873632.1 8 - 9 1190 105
XM_017018144.1 XP_016873633.1 8 - 9 1092 105
XM_017018145.1 XP_016873634.1 7 - 8 3124 105
XM_017018146.1 XP_016873635.1 8 - 9 1254 105
XM_017018147.1 XP_016873636.1 7 - 8 3051 105
XM_017018148.1 XP_016873637.1 6 - 7 2983 105
XM_017018149.1 XP_016873638.1 7 - 8 1135 105
XM_017018150.1 XP_016873639.1 7 - 8 990 105
XM_017018151.1 XP_016873640.1 7 - 8 949 105
XM_017018152.1 XP_016873641.1 5 - 6 1280 105
XM_017018153.1 XP_016873642.1 5 - 6 878 105
XM_017018154.1 XP_016873643.1 5 - 6 737 105
XM_017018155.1 XP_016873644.1 5 - 6 1292 105
XM_017018156.1 XP_016873645.1 5 - 6 738 105
XM_017018157.1 XP_016873646.1 5 - 6 503 105
XM_017018158.1 XP_016873647.1 6 - 7 789 105
XM_017018159.1 XP_016873648.1 7 - 8 3127 105
XM_017018160.1 XP_016873649.1 5 - 6 926 105
XM_017018161.1 XP_016873650.1 7 - 8 3133 105
XM_017018162.1 XP_016873651.1 6 - 7 736 105
XM_017018163.1 XP_016873652.1 5 - 6 727 105
XM_017018164.1 XP_016873653.1 5 - 6 613 105
XM_017018165.1 XP_016873654.1 6 - 7 692 105
GenBank mRNA AJ303380.2 - 4 - 5 1477 105
AK092720.1 - 3 - 4 365 105
AK093251.1 - 5 - 6 842 105
AK122808.1 - 2 - 3 3052 105
AK293193.1 - 6 - 7 585 105
BC031231.1 - 6 - 7 1081 105

View Full Product Details