The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012162.3 | NP_036294.2 | 2 - 3 | 650 | 100 |
GenBank mRNA | AF174592.1 | - | 2 - 3 | 307 | 100 |
AF370422.1 | - | 2 - 3 | 1646 | 100 | |
BC033066.1 | - | 2 - 3 | 639 | 100 |