Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXL6
Entrez Gene ID
26233
Gene Name
F-box and leucine rich repeat protein 6
Gene Aliases
FBL6, FBL6A, PP14630
Chromosome Location
Chr.8: 144355428 - 144358523 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012162.3 NP_036294.2 4 - 5 849 79
NM_024555.5 NP_078831.4 4 - 5 831 79
GenBank mRNA AF174592.1 - 4 - 5 506 79
AF370422.1 - 3 - 4 2162 79
AK026541.1 - 4 - 5 780 79
AK223088.1 - 4 - 5 780 79
BC033066.1 - 4 - 5 838 79

View Full Product Details