The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006668.1 | NP_006659.1 | 3 - 4 | 275 | 77 |
XM_011536364.1 | XP_011534666.1 | 3 - 4 | 431 | 77 | |
GenBank mRNA | AF094480.1 | - | 3 - 4 | 275 | 77 |
AK295216.1 | - | 3 - 4 | 237 | 77 | |
BC022539.1 | - | 3 - 4 | 376 | 77 |