The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004958.3 | NP_004949.1 | 6 - 7 | 960 | 63 |
XM_005263438.2 | XP_005263495.1 | 6 - 7 | 967 | 63 | |
XM_011541166.2 | XP_011539468.1 | 6 - 7 | 960 | 63 | |
XM_017000900.1 | XP_016856389.1 | 3 - 4 | 423 | 63 | |
XM_017000901.1 | XP_016856390.1 | 5 - 6 | 820 | 63 | |
XM_017000902.1 | XP_016856391.1 | 6 - 7 | 960 | 63 | |
GenBank mRNA | AB209995.1 | - | 6 - 7 | 941 | 63 |
AK302863.1 | - | 6 - 7 | 952 | 63 | |
AK304273.1 | - | 6 - 7 | 949 | 63 | |
BC117166.1 | - | 5 - 6 | 857 | 63 | |
L34075.1 | - | 6 - 7 | 918 | 63 | |
U88966.1 | - | 6 - 7 | 918 | 63 |