The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173595.3 | NP_775866.2 | 18 - 19 | 2071 | 68 |
XM_011538197.2 | XP_011536499.1 | 17 - 18 | 2173 | 68 | |
XM_017019183.1 | XP_016874672.1 | 17 - 18 | 2254 | 68 | |
XM_017019184.1 | XP_016874673.1 | 18 - 19 | 2103 | 68 | |
GenBank mRNA | AB190990.1 | - | 18 - 19 | 2088 | 68 |
AK125379.1 | - | 18 - 19 | 2088 | 68 | |
AK126893.1 | - | 11 - 12 | 1231 | 68 |