Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOLT1B
Entrez Gene ID
51026
Gene Name
golgi transport 1B
Gene Aliases
CGI-141, GCT2, GOT1, GOT1B, YMR292W
Chromosome Location
Chr.12: 21501765 - 21518403 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016072.4 NP_057156.1 1 - 2 186 119
XM_017019384.1 XP_016874873.1 1 - 2 170 119
GenBank mRNA AB097020.1 - 1 - 2 136 119
AF151899.1 - 1 - 2 120 119
AK310889.1 - 1 - 2 122 119
AK311920.1 - 1 - 2 98 119
AL136571.1 - 1 - 2 85 119
AY358975.1 - 1 - 2 103 119
BC012455.1 - 1 - 2 107 119
DA579356.1 - 1 - 2 186 119

View Full Product Details