The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003000.2 | NP_002991.2 | 5 - 6 | 689 | 77 |
GenBank mRNA | AK312056.1 | - | 5 - 6 | 551 | 77 |
BC007840.2 | - | 5 - 6 | 689 | 77 | |
BF796275.1 | - | 5 - 6 | 537 | 77 | |
BU554163.1 | - | 3 - 4 | 279 | 77 | |
D10245.1 | - | 4 - 5 | 484 | 77 | |
DQ403007.1 | - | 5 - 6 | 511 | 77 | |
M32246.1 | - | 4 - 5 | 484 | 77 | |
U17248.1 | - | 5 - 6 | 672 | 77 |