Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGBL2
Entrez Gene ID
79841
Gene Name
ATP/GTP binding protein like 2
Gene Aliases
CCP2
Chromosome Location
Chr.11: 47659168 - 47715397 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024783.3 NP_079059.2 11 - 12 2085 109
XM_005253138.4 XP_005253195.1 9 - 10 2416 109
XM_005253139.4 XP_005253196.1 9 - 10 2416 109
XM_005253140.4 XP_005253197.1 9 - 10 2416 109
XM_006718327.3 XP_006718390.1 8 - 9 2362 109
XM_006718328.3 XP_006718391.1 8 - 9 2362 109
XM_011520372.2 XP_011518674.1 10 - 11 2325 109
XM_011520373.2 XP_011518675.1 9 - 10 2416 109
XM_011520374.2 XP_011518676.1 8 - 9 2308 109
XM_011520375.2 XP_011518677.1 8 - 9 2281 109
XM_011520376.2 XP_011518678.1 10 - 11 2621 109
XM_011520377.2 XP_011518679.1 7 - 8 1536 109
XM_011520378.2 XP_011518680.1 10 - 11 1886 109
XM_011520379.2 XP_011518681.1 9 - 10 1681 109
XM_011520381.2 XP_011518683.1 9 - 10 2416 109
XM_017018331.1 XP_016873820.1 9 - 10 2416 109
XM_017018332.1 XP_016873821.1 7 - 8 1585 109
GenBank mRNA AK027251.1 - 2 - 3 742 109
AK056687.1 - 8 - 9 2315 109
AK223171.1 - 2 - 3 742 109
AK223176.1 - 2 - 3 742 109
AK303066.1 - 10 - 11 1924 109
BC036234.1 - 11 - 12 2069 109

View Full Product Details