Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC6A9
Entrez Gene ID
6536
Gene Name
solute carrier family 6 member 9
Gene Aliases
GLYT1
Chromosome Location
Chr.1: 43996483 - 44031492 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024845.2 NP_001020016.1 12 - 13 1805 68
NM_001261380.1 NP_001248309.1 11 - 12 1752 68
NM_006934.3 NP_008865.2 11 - 12 1797 68
NM_201649.3 NP_964012.2 12 - 13 1959 68
NR_048548.1 11 - 12 1793 68
XM_017002151.1 XP_016857640.1 12 - 13 1713 68
XM_017002152.1 XP_016857641.1 11 - 12 1687 68
XM_017002153.1 XP_016857642.1 11 - 12 1458 68
GenBank mRNA AK296438.1 - 11 - 12 1555 68
AK298611.1 - 10 - 11 1410 68
AK315233.1 - 11 - 12 1796 68
S70609.1 - 12 - 13 1943 68
S70612.1 - 11 - 12 1781 68

View Full Product Details