The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017421.3 | NP_059117.3 | 6 - 7 | 919 | 99 |
XM_005267018.4 | XP_005267075.1 | 5 - 6 | 692 | 99 | |
XM_006715500.3 | XP_006715563.1 | 7 - 8 | 1058 | 99 | |
XM_011535898.2 | XP_011534200.1 | 6 - 7 | 816 | 99 | |
XM_011535899.1 | XP_011534201.1 | 5 - 6 | 747 | 99 | |
XM_017010933.1 | XP_016866422.1 | 7 - 8 | 916 | 99 | |
GenBank mRNA | AF193016.1 | - | 6 - 7 | 856 | 99 |
AK056955.1 | - | 6 - 7 | 917 | 99 | |
AL136726.1 | - | 6 - 7 | 917 | 99 | |
BC001534.1 | - | 6 - 7 | 890 | 99 | |
BC015634.2 | - | 6 - 7 | 880 | 99 | |
BC063463.1 | - | 6 - 7 | 895 | 99 |