The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320559.1 | NP_001307488.1 | 3 - 4 | 817 | 111 |
NM_001320560.1 | NP_001307489.1 | 3 - 4 | 618 | 111 | |
NM_033027.3 | NP_149016.2 | 3 - 4 | 686 | 111 | |
XM_017007048.1 | XP_016862537.1 | 3 - 4 | 3782 | 111 | |
GenBank mRNA | AB053121.1 | - | 3 - 4 | 663 | 111 |
AB063301.1 | - | 3 - 4 | 578 | 111 | |
BC038949.1 | - | 3 - 4 | 633 | 111 | |
BX397882.2 | - | 3 - 4 | 618 | 110 |