The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174910.1 | NP_777570.1 | 1 - 2 | 200 | 101 |
XM_006715554.3 | XP_006715617.1 | 1 - 2 | 239 | 101 | |
XM_011536093.2 | XP_011534395.1 | 1 - 2 | 239 | 101 | |
GenBank mRNA | AF519569.1 | - | 1 - 2 | 200 | 101 |
BC113639.1 | - | 1 - 2 | 141 | 101 |