Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCTE3
Entrez Gene ID
6991
Gene Name
t-complex-associated-testis-expressed 3
Gene Aliases
TCTEX1D3
Chromosome Location
Chr.6: 169725488 - 169751581 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174910.1 NP_777570.1 2 - 3 406 120
XM_011536093.2 XP_011534395.1 2 - 3 445 120
GenBank mRNA AF519569.1 - 2 - 3 406 120
BC113639.1 - 2 - 3 347 120

View Full Product Details