The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003865.2 | NP_003856.1 | 2 - 3 | 692 | 86 |
XM_005265526.4 | XP_005265583.1 | 3 - 4 | 1816 | 86 | |
XM_006713379.3 | XP_006713442.1 | 3 - 4 | 586 | 86 | |
XM_011534204.2 | XP_011532506.1 | 4 - 5 | 1341 | 86 | |
XM_011534205.2 | XP_011532507.1 | 4 - 5 | 612 | 86 | |
XM_017007421.1 | XP_016862910.1 | 4 - 5 | 676 | 86 | |
GenBank mRNA | BC069515.1 | - | 2 - 3 | 427 | 86 |
BC093979.1 | - | 2 - 3 | 404 | 86 | |
BC112089.1 | - | 2 - 3 | 404 | 86 |