The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003842.4 | NP_003833.4 | 7 - 8 | 1225 | 86 |
NM_147187.2 | NP_671716.2 | 8 - 9 | 1138 | 86 | |
NR_027140.1 | 7 - 8 | 1032 | 86 | ||
XM_017013944.1 | XP_016869433.1 | 7 - 8 | 1013 | 86 | |
GenBank mRNA | AB451272.1 | - | 7 - 8 | 932 | 86 |
AB451399.1 | - | 7 - 8 | 932 | 86 | |
AF012535.1 | - | 8 - 9 | 984 | 86 | |
AF012628.1 | - | 8 - 9 | 845 | 86 | |
AF016266.1 | - | 7 - 8 | 1049 | 86 | |
AF016268.1 | - | 8 - 9 | 954 | 86 | |
AF016849.1 | - | 7 - 8 | 1076 | 86 | |
AF018657.1 | - | 8 - 9 | 845 | 86 | |
AF018658.1 | - | 7 - 8 | 932 | 86 | |
AF020501.1 | - | 8 - 9 | 845 | 86 | |
AF022386.1 | - | 8 - 9 | 845 | 86 | |
AF153687.1 | - | 8 - 9 | 963 | 86 | |
AF192548.1 | - | 8 - 9 | 945 | 86 | |
AK291216.1 | - | 7 - 8 | 1052 | 86 | |
AK296128.1 | - | 3 - 4 | 238 | 86 | |
AK298602.1 | - | 7 - 8 | 876 | 86 | |
AY358277.1 | - | 8 - 9 | 984 | 86 | |
BC001281.1 | - | 7 - 8 | 1079 | 86 | |
BX537617.1 | - | 2 - 3 | 330 | 86 | |
BX538104.1 | - | 7 - 8 | 1054 | 86 | |
BX538179.1 | - | 2 - 3 | 330 | 86 | |
CR541898.1 | - | 7 - 8 | 932 | 86 | |
CX870630.1 | - | 3 - 4 | 437 | 86 | |
FJ200482.1 | - | 6 - 7 | 826 | 86 |