The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030582.3 | NP_085059.2 | 21 - 22 | 2720 | 152 |
NM_130444.2 | NP_569711.2 | 21 - 22 | 3425 | 152 | |
NM_130445.3 | NP_569712.2 | 22 - 23 | 2253 | 152 | |
GenBank mRNA | AF018081.1 | - | 21 - 22 | 2739 | 152 |
AF018082.1 | - | 22 - 23 | 2218 | 152 | |
BC033715.1 | - | 10 - 11 | 601 | 152 | |
BC063833.1 | - | 22 - 23 | 2194 | 152 | |
L22548.1 | - | 3 - 4 | 200 | 152 |