The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030582.3 | NP_085059.2 | 9 - 10 | 1876 | 164 |
NM_130444.2 | NP_569711.2 | 9 - 10 | 2581 | 164 | |
NM_130445.3 | NP_569712.2 | 10 - 11 | 1409 | 164 | |
GenBank mRNA | AF018081.1 | - | 9 - 10 | 164 | |
AF018082.1 | - | 10 - 11 | 164 | ||
BC063833.1 | - | 10 - 11 | 1350 | 164 |