Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C2orf76
Entrez Gene ID
130355
Gene Name
chromosome 2 open reading frame 76
Gene Aliases
AIM29
Chromosome Location
Chr.2: 119281472 - 119366879 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017927.3 NP_001017927.2 5 - 6 830 113
NM_001322329.1 NP_001309258.1 5 - 6 751 113
NM_001322330.1 NP_001309259.1 4 - 5 627 113
NM_001322331.1 NP_001309260.1 4 - 5 280 113
XM_017003354.1 XP_016858843.1 4 - 5 329 113
GenBank mRNA BC047365.1 - 5 - 6 751 113
BC062434.1 - 3 - 4 262 113
BC105805.1 - 4 - 5 284 113
BC126395.1 - 4 - 5 567 113
BC126397.1 - 5 - 6 770 113
BC144012.1 - 5 - 6 770 113
BC171737.1 - 4 - 5 567 113

View Full Product Details