Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTOA
Entrez Gene ID
146183
Gene Name
otoancorin
Gene Aliases
CT108, DFNB22
Chromosome Location
Chr.16: 21663857 - 21771468 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161683.1 NP_001155155.1 7 - 8 960 93
NM_144672.3 NP_653273.3 11 - 12 1104 93
NM_170664.2 NP_733764.1 2 - 3 198 93
XM_011545747.2 XP_011544049.1 12 - 13 1483 93
XM_017022950.1 XP_016878439.1 12 - 13 1207 93
GenBank mRNA AK093062.1 - 2 - 3 198 93
AK125840.1 - 7 - 8 960 93
AK302733.1 - 1 - 2 260 93
BC129992.1 - 11 - 12 1101 93
BC129993.1 - 11 - 12 1108 93

View Full Product Details