The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161683.1 | NP_001155155.1 | 8 - 9 | 1179 | 74 |
NM_144672.3 | NP_653273.3 | 12 - 13 | 1323 | 74 | |
NM_170664.2 | NP_733764.1 | 3 - 4 | 417 | 74 | |
XM_011545747.2 | XP_011544049.1 | 13 - 14 | 1702 | 74 | |
XM_011545748.2 | XP_011544050.2 | 2 - 3 | 696 | 74 | |
XM_017022950.1 | XP_016878439.1 | 13 - 14 | 1426 | 74 | |
GenBank mRNA | AK093062.1 | - | 3 - 4 | 417 | 74 |
AK125840.1 | - | 8 - 9 | 1179 | 74 | |
AK302733.1 | - | 2 - 3 | 479 | 74 | |
BC129992.1 | - | 12 - 13 | 1320 | 74 | |
BC129993.1 | - | 12 - 13 | 1327 | 74 |