Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTOA
Entrez Gene ID
146183
Gene Name
otoancorin
Gene Aliases
CT108, DFNB22
Chromosome Location
Chr.16: 21663857 - 21771468 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161683.1 NP_001155155.1 9 - 10 1343 79
NM_144672.3 NP_653273.3 13 - 14 1487 79
NM_170664.2 NP_733764.1 4 - 5 581 79
XM_011545747.2 XP_011544049.1 14 - 15 1866 79
XM_011545748.2 XP_011544050.2 3 - 4 860 79
XM_017022950.1 XP_016878439.1 14 - 15 1590 79
GenBank mRNA AK093062.1 - 4 - 5 581 79
AK125840.1 - 9 - 10 1343 79
AK302733.1 - 3 - 4 643 79
BC129992.1 - 13 - 14 1484 79
BC129993.1 - 13 - 14 1491 79

View Full Product Details