Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OTOA
Entrez Gene ID
146183
Gene Name
otoancorin
Gene Aliases
CT108, DFNB22
Chromosome Location
Chr.16: 21663857 - 21771468 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161683.1 NP_001155155.1 11 - 12 1550 90
NM_144672.3 NP_653273.3 15 - 16 1694 90
NM_170664.2 NP_733764.1 6 - 7 788 90
XM_011545747.2 XP_011544049.1 16 - 17 2073 90
XM_011545748.2 XP_011544050.2 5 - 6 1067 90
XM_017022950.1 XP_016878439.1 16 - 17 1797 90
GenBank mRNA AK093062.1 - 6 - 7 788 90
AK125840.1 - 11 - 12 1550 90
AK302733.1 - 5 - 6 850 90
BC129992.1 - 15 - 16 1691 90
BC129993.1 - 15 - 16 1698 90

View Full Product Details