The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005562.2 | NP_005553.2 | 21 - 22 | 3538 | 75 |
NM_018891.2 | NP_061486.2 | 21 - 22 | 3538 | 75 | |
GenBank mRNA | BC112286.1 | - | 21 - 22 | 3265 | 75 |
BC113378.1 | - | 21 - 22 | 3265 | 75 | |
BC143653.1 | - | 21 - 22 | 3242 | 75 | |
X73902.1 | - | 21 - 22 | 3289 | 75 | |
Z15008.1 | - | 21 - 22 | 3341 | 75 | |
Z15009.1 | - | 21 - 22 | 3341 | 75 |