The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113347.1 | NP_001106818.1 | 5 - 6 | 1050 | 78 |
NM_001113348.1 | NP_001106819.1 | 7 - 8 | 945 | 78 | |
NM_001113349.1 | NP_001106820.1 | 6 - 7 | 896 | 78 | |
NM_001397.2 | NP_001388.1 | 7 - 8 | 905 | 78 | |
XM_006710398.2 | XP_006710461.1 | 7 - 8 | 1050 | 78 | |
XM_011540872.2 | XP_011539174.1 | 7 - 8 | 1657 | 78 | |
XM_011540873.2 | XP_011539175.1 | 7 - 8 | 975 | 78 | |
XM_017000511.1 | XP_016856000.1 | 7 - 8 | 797 | 78 | |
GenBank mRNA | AB031742.1 | - | 6 - 7 | 790 | 78 |
AK290656.1 | - | 7 - 8 | 877 | 78 | |
AK296486.1 | - | 7 - 8 | 880 | 78 | |
AK304167.1 | - | 6 - 7 | 838 | 78 | |
AK309677.1 | - | 7 - 8 | 879 | 78 | |
BC117256.1 | - | 7 - 8 | 905 | 78 | |
BC126257.1 | - | 7 - 8 | 905 | 78 | |
BX648351.1 | - | 5 - 6 | 909 | 78 | |
D43698.1 | - | 5 - 6 | 855 | 78 | |
D49471.1 | - | 7 - 8 | 830 | 78 | |
DA445355.1 | - | 3 - 4 | 349 | 78 | |
Z35307.1 | - | 7 - 8 | 816 | 78 |