The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004714.2 | NP_004705.1 | 3 - 4 | 489 | 68 |
NM_006483.2 | NP_006474.1 | 3 - 4 | 489 | 68 | |
NM_006484.2 | NP_006475.1 | 3 - 4 | 489 | 68 | |
XM_005259398.4 | XP_005259455.1 | 3 - 4 | 941 | 68 | |
GenBank mRNA | AF205861.1 | - | 3 - 4 | 386 | 68 |
AK074971.1 | - | 3 - 4 | 405 | 68 | |
AK302127.1 | - | 3 - 4 | 418 | 68 | |
BC018751.1 | - | 3 - 4 | 441 | 68 | |
BC025291.1 | - | 3 - 4 | 392 | 68 | |
Y17999.1 | - | 3 - 4 | 457 | 68 |