The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142782.1 | NP_001136254.1 | 8 - 9 | 1346 | 97 |
NM_152900.2 | NP_690864.2 | 8 - 9 | 1346 | 97 | |
XM_005270737.3 | XP_005270794.1 | 8 - 9 | 1346 | 97 | |
XM_011541208.2 | XP_011539510.1 | 8 - 9 | 3297 | 97 | |
XM_017000974.1 | XP_016856463.1 | 8 - 9 | 1725 | 97 | |
XM_017000975.1 | XP_016856464.1 | 2 - 3 | 318 | 97 | |
GenBank mRNA | AB046854.1 | - | 4 - 5 | 412 | 97 |
AF213259.1 | - | 8 - 9 | 1165 | 97 | |
BC130409.1 | - | 8 - 9 | 1346 | 97 |