Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DHRS3
Entrez Gene ID
9249
Gene Name
dehydrogenase/reductase 3
Gene Aliases
DD83.1, RDH17, Rsdr1, SDR1, SDR16C1, retSDR1
Chromosome Location
Chr.1: 12567910 - 12618361 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319225.1 NP_001306154.1 3 - 4 491 79
NM_001324370.1 NP_001311299.1 3 - 4 454 79
NM_004753.6 NP_004744.2 3 - 4 844 79
XM_017002809.1 XP_016858298.1 3 - 4 369 79
GenBank mRNA AF061741.1 - 3 - 4 515 79
AK222593.1 - 3 - 4 515 79
AK225057.1 - 3 - 4 491 79
AK312961.1 - 3 - 4 799 79
BC002730.2 - 3 - 4 576 79
BF037805.1 - - 152 79
BX648476.1 - 3 - 4 850 79

View Full Product Details