The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001036.4 | NP_001027.3 | 20 - 21 | 2661 | 73 |
NM_001243996.2 | NP_001230925.1 | 20 - 21 | 2661 | 73 | |
XM_011521880.2 | XP_011520182.1 | 20 - 21 | 2849 | 73 | |
XM_017022468.1 | XP_016877957.1 | 20 - 21 | 2849 | 73 | |
XM_017022469.1 | XP_016877958.1 | 20 - 21 | 2849 | 73 | |
XM_017022470.1 | XP_016877959.1 | 20 - 21 | 5183 | 73 | |
XM_017022471.1 | XP_016877960.1 | 20 - 21 | 2849 | 73 | |
XM_017022472.1 | XP_016877961.1 | 20 - 21 | 2849 | 73 | |
XM_017022473.1 | XP_016877962.1 | 20 - 21 | 2849 | 73 | |
XM_017022474.1 | XP_016877963.1 | 20 - 21 | 2813 | 73 | |
XM_017022475.1 | XP_016877964.1 | 20 - 21 | 2849 | 73 | |
XM_017022476.1 | XP_016877965.1 | 19 - 20 | 2741 | 73 | |
XM_017022477.1 | XP_016877966.1 | 20 - 21 | 2850 | 73 | |
GenBank mRNA | AB001025.1 | - | 20 - 21 | 2666 | 73 |
AJ001515.1 | - | 20 - 21 | 2647 | 73 |