The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001036.4 | NP_001027.3 | 35 - 36 | 5495 | 73 |
NM_001243996.2 | NP_001230925.1 | 35 - 36 | 5495 | 73 | |
XM_011521880.2 | XP_011520182.1 | 35 - 36 | 5683 | 73 | |
XM_017022468.1 | XP_016877957.1 | 35 - 36 | 5683 | 73 | |
XM_017022469.1 | XP_016877958.1 | 35 - 36 | 5683 | 73 | |
XM_017022470.1 | XP_016877959.1 | 35 - 36 | 8017 | 73 | |
XM_017022471.1 | XP_016877960.1 | 35 - 36 | 5680 | 73 | |
XM_017022472.1 | XP_016877961.1 | 35 - 36 | 5683 | 73 | |
XM_017022473.1 | XP_016877962.1 | 35 - 36 | 5683 | 73 | |
XM_017022474.1 | XP_016877963.1 | 35 - 36 | 5647 | 73 | |
XM_017022475.1 | XP_016877964.1 | 35 - 36 | 5683 | 73 | |
XM_017022476.1 | XP_016877965.1 | 34 - 35 | 5575 | 73 | |
XM_017022477.1 | XP_016877966.1 | 35 - 36 | 5684 | 73 | |
GenBank mRNA | AB001025.1 | - | 35 - 36 | 5500 | 73 |
AJ001515.1 | - | 35 - 36 | 5481 | 73 |