Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PPP1R9A
- Entrez Gene ID
-
55607
- Gene Name
- protein phosphatase 1 regulatory subunit 9A
- Gene Aliases
-
NRB1, NRBI, Neurabin-I
- Chromosome Location
-
Chr.7: 94907202 - 95296415 on Build GRCh38
View Full Product Details