The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166161.1 | NP_001159633.1 | 15 - 16 | 3584 | 89 |
NM_001166162.1 | NP_001159634.1 | 15 - 16 | 3638 | 89 | |
XM_017012395.1 | XP_016867884.1 | 18 - 19 | 3833 | 89 | |
XM_017012397.1 | XP_016867886.1 | 18 - 19 | 3833 | 89 | |
XM_017012400.1 | XP_016867889.1 | 16 - 17 | 3704 | 89 | |
XM_017012401.1 | XP_016867890.1 | 17 - 18 | 3716 | 89 | |
XM_017012402.1 | XP_016867891.1 | 16 - 17 | 3650 | 89 | |
XM_017012403.1 | XP_016867892.1 | 16 - 17 | 3650 | 89 | |
GenBank mRNA | AK000075.1 | - | 5 - 6 | 715 | 89 |
AK225204.1 | - | 5 - 6 | 715 | 89 | |
BC130449.1 | - | 15 - 16 | 3453 | 89 | |
BC144108.1 | - | 15 - 16 | 3401 | 89 |