The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017839.4 | NP_060309.2 | 7 - 8 | 974 | 71 |
XM_005256006.2 | XP_005256063.1 | 7 - 8 | 903 | 71 | |
XM_011523169.2 | XP_011521471.1 | 7 - 8 | 874 | 71 | |
XM_017023376.1 | XP_016878865.1 | 8 - 9 | 948 | 71 | |
GenBank mRNA | AB244718.1 | - | 7 - 8 | 793 | 71 |
AK000488.1 | - | 7 - 8 | 901 | 71 | |
AK308386.1 | - | 7 - 8 | 902 | 71 | |
BC002472.2 | - | 7 - 8 | 933 | 71 | |
BX641069.1 | - | 2 - 3 | 875 | 71 |