Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDGFC
Entrez Gene ID
56034
Gene Name
platelet derived growth factor C
Gene Aliases
FALLOTEIN, SCDGF
Chromosome Location
Chr.4: 156761611 - 156971394 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016205.2 NP_057289.1 4 - 5 1192 94
NR_036641.1 5 - 6 1253 94
XM_011532124.2 XP_011530426.1 6 - 7 872 94
XM_017008455.1 XP_016863944.1 4 - 5 821 94
GenBank mRNA AB033831.1 - 4 - 5 1027 94
AF091434.1 - 4 - 5 1192 94
AF244813.1 - 4 - 5 889 94
AF260738.1 - 4 - 5 863 94
AK304638.1 - 6 - 7 535 94
AK313817.1 - 4 - 5 922 94
AM922296.1 - 5 - 6 1073 94
AY358493.1 - 4 - 5 985 94
BC094746.1 - 4 - 5 793 94
BC136662.1 - 4 - 5 1113 94

View Full Product Details