Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC15
Entrez Gene ID
79770
Gene Name
thioredoxin domain containing 15
Gene Aliases
C5orf14, UNQ335
Chromosome Location
Chr.5: 134873770 - 134901633 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024715.3 NP_078991.3 3 - 4 1413 77
GenBank mRNA AK026278.1 - 4 - 5 816 77
AK056512.1 - 3 - 4 1448 77
AK092425.1 - 1 - 2 1478 77
AK225454.1 - 4 - 5 816 77
AK225462.1 - 4 - 5 816 77
AK299578.1 - 3 - 4 706 77
AK307529.1 - 2 - 3 324 77
AY358649.1 - 3 - 4 841 77
BC001615.1 - 3 - 4 900 77
BC032568.1 - 3 - 4 814 77
BX538131.1 - 3 - 4 740 77

View Full Product Details