Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC15
Entrez Gene ID
79770
Gene Name
thioredoxin domain containing 15
Gene Aliases
C5orf14, UNQ335
Chromosome Location
Chr.5: 134873770 - 134901633 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024715.3 NP_078991.3 4 - 5 1545 84
GenBank mRNA AK026278.1 - 5 - 6 948 84
AK056512.1 - 4 - 5 1580 84
AK092425.1 - 2 - 3 1610 84
AK225454.1 - 5 - 6 948 84
AK225462.1 - 5 - 6 948 84
AK299578.1 - 4 - 5 838 84
AK307529.1 - 3 - 4 456 84
AY358649.1 - 4 - 5 973 84
BC001615.1 - 4 - 5 1032 84
BC032568.1 - 4 - 5 946 84
BX538131.1 - 4 - 5 872 84

View Full Product Details