Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CENPK
Entrez Gene ID
64105
Gene Name
centromere protein K
Gene Aliases
AF5alpha, CENP-K, FKSG14, P33, Solt
Chromosome Location
Chr.5: 65488919 - 65563194 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267038.1 NP_001253967.1 10 - 11 822 166
NM_022145.4 NP_071428.2 10 - 11 856 166
XM_011543534.2 XP_011541836.1 10 - 11 800 166
XM_011543535.2 XP_011541837.1 10 - 11 918 166
XM_011543536.2 XP_011541838.1 11 - 12 1078 166
XM_011543537.2 XP_011541839.1 10 - 11 834 166
XM_011543538.2 XP_011541840.1 9 - 10 701 166
XM_011543539.2 XP_011541841.1 9 - 10 783 166
XM_011543541.2 XP_011541843.1 7 - 8 602 166
XM_011543542.2 XP_011541844.1 7 - 8 722 166
XM_017009690.1 XP_016865179.1 10 - 11 1061 166
XM_017009691.1 XP_016865180.1 10 - 11 812 166
XM_017009692.1 XP_016865181.1 10 - 11 1030 166
XM_017009694.1 XP_016865183.1 11 - 12 1029 166
XM_017009695.1 XP_016865184.1 9 - 10 698 166
XM_017009697.1 XP_016865186.1 9 - 10 732 166
XM_017009698.1 XP_016865187.1 8 - 9 652 166
XM_017009700.1 XP_016865189.1 9 - 10 3166 166
XM_017009701.1 XP_016865190.1 8 - 9 797 166
GenBank mRNA AF315941.1 - 10 - 11 812 166
AY009151.1 - 8 - 9 686 166
BC005400.1 - 10 - 11 812 166
BC008504.1 - 10 - 11 856 166

View Full Product Details