The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152491.4 | NP_689704.4 | 7 - 8 | 979 | 96 |
XM_017000396.1 | XP_016855885.1 | 7 - 8 | 973 | 96 | |
XM_017000398.1 | XP_016855887.1 | 7 - 8 | 973 | 96 | |
GenBank mRNA | AK290786.1 | - | 7 - 8 | 963 | 96 |
BC039170.1 | - | 7 - 8 | 948 | 96 | |
BC063477.1 | - | 7 - 8 | 948 | 96 |